Learn More
SMCC-DM1, TRC
Specifications
Specifications
| Chemical Name or Material | SMCC-DM1 |
| CAS | 1228105-51-8 |
| Molecular Formula | C26 H28 O13 |
| Quantity | 5 mg |
| Synonym | 8-[(2S,3R,4R,5S,6R)-6-[[(2R,3R,4R)-3,4-dihydroxy-4-(hydroxymethyl)oxolan-2-yl]oxymethyl]-3,4,5-trihydroxyoxan-2-yl]-7-hydroxy-3-(4-hydroxyphenyl)chromen-4-one, SMCC-DM1 |
| InChI Formula | InChI=1S/C26H28O13/c27-9-26(35)10-38-25(24(26)34)37-8-16-19(31)20(32)21(33)23(39-16)17-15(29)6-5-13-18(30)14(7-36-22(13)17)11-1-3-12(28)4-2-11/h1-7,16,19-21,23-25,27-29,31-35H,8-10H2/t16-,19-,20+,21-,23+,24+,25-,26-/m1/s1 |
| SMILES | OC[C@@]1(O)CO[C@@H](OC[C@H]2O[C@H]([C@H](O)[C@@H](O)[C@@H]2O)c3c(O)ccc4C(=O)C(=COc34)c5ccc(O)cc5)[C@@H]1O |
| IUPAC Name | 8-[(2S,3R,4R,5S,6R)-6-[[(2R,3R,4R)-3,4-dihydroxy-4-(hydroxymethyl)oxolan-2-yl]oxymethyl]-3,4,5-trihydroxyoxan-2-yl]-7-hydroxy-3-(4-hydroxyphenyl)chromen-4-one |
| Molecular Weight (g/mol) | 548.493 |
| Formula Weight | 548.153 |