Learn More
Abnova™ SMARCB1/CEN22q FISH Probe
Labeled FISH probes for identification of gene amplification using Fluoresecent In Situ Hybridization Technique.
Brand:  Abnova™ FG0208.100uL
Additional Details : Weight : 0.02000kg
 View more versions of this product
Product Code. 16043335
  • 1300.00 € / 100µL
Estimated Shipment: 10-11-2023
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Specifications
6598
BAF47,INI1,RDT,SNF5,SNF5L1,Sfh1p,Snr1,hSNFS
Fluorescent In Situ Hybridization (Cell)
100 μL
SMARCB1
Store at 4°C in the dark
SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily b, member 1
DNA/RNA