missing translation for 'onlineSavingsMsg'
Learn More

Smad9 [p Tyr92] Antibody (ARC65536) [Alexa Fluor« 700], Novus Biologicals Biologicals™

Product Code. 30500735 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30500735 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30500735 Supplier Novus Biologicals Supplier No. NBP333580AF700

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Smad9 Monoclonal antibody specifically detects Smad9 in Human samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Smad9
Applications ELISA, Western Blot
Classification Monoclonal
Clone ARC65536
Conjugate Alexa Fluor 700
Formulation 50mM Sodium Borate
Gene Alias MAD homolog 9, MAD, mothers against decapentaplegic homolog 9 (Drosophila), Madh6, MADH6mothers against decapentaplegic homolog 9, MADH9Mothers against decapentaplegic, drosophila, homolog of, 9, Mothers against DPP homolog 9, SMAD 9, SMAD family member 9SMAD8B, SMAD, mothers against DPP homolog 9, SMAD, mothers against DPP homolog 9 (Drosophila), SMAD8, SMAD8A, Smad9
Host Species Rabbit
Immunogen A synthetic phosphorylated peptide around Y92 of human Smad9.1).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4093
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.