missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Smad7 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
285.00 € - 539.00 €
Specifications
| Antigen | Smad7 |
|---|---|
| Dilution | Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:50-1:200 |
| Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18448261
|
Novus Biologicals
NBP1-87728-25ul |
25 μL |
285.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18239246
|
Novus Biologicals
NBP1-87728 |
0.1 mL |
539.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Smad7 Polyclonal specifically detects Smad7 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| Smad7 | |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 4092 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:EVKRLCCCESYGKINPELVCCNPHHLSRLCELESPPPPYSRYPMDFLKPTADCPDAVPSSAETGGTNYLAPGGLSDSQLLLEPGDRSHWCVVAYWEEKTRVGRLYCVQEPSLDIFYDLPQGNG | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:50-1:200 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| CRCS3, FLJ16482, hSMAD7, MAD homolog 7, MAD homolog 8, MAD, mothers against decapentaplegic homolog 7 (Drosophila), MADH7mothers against decapentaplegic homolog 7, MADH8, Mothers against decapentaplegic homolog 8, Mothers against decapentaplegic, drosophila, homolog of, 7, Mothers against DPP homolog 7, Mothers against DPP homolog 8, SMAD 7, SMAD family member 7MAD (mothers against decapentaplegic, Drosophila) homolog 7, SMAD, mothers against DPP homolog 7, SMAD, mothers against DPP homolog 7 (Drosophila), Smad7 | |
| SMAD7 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title