missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ SM 324405

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
SM 324405 is a potent TLR7 agonist (EC50 = 50 nM). Exhibits selectivity for TLR7 over TLR8. Induces IFN-α and -γ expression in human PBMCs and mouse splenocytes, and inhibits IL-5 production in human PBMCs in vitro. Rapidly metabolized to acid metabolite (t½ = 2.6 min in human plasma).
Specifications
Specifications
| Chemical Name or Material | Methyl 3-[(6-amino-2-butoxy-7,8-dihydro-8-oxo-9H-purin-9-yl)methyl]benzeneacetate |
| CAS | 677773-91-0 |
| Quantity | 10 mg |
| Target | Toll-like Receptor Agonists |
| Molecular Formula | C19H23N5O4 |
| Formula Weight | 385.42 |
| Purity | >98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction