Learn More
Edmund Bühler GmbH™ Sliding Plate with Combifix SM
Affix to a multistory platform. Edmund Bühler GmbH™ Sliding Plate with Combifix SM fastens horizontal separating funnels of different sizes to platform.
Brand: Edmund Bühler GmbH™ 0051 486
Additional Details : Weight : 0.50000kg
Description
- Fastens horizontal separating funnels of different sizes to platform
- For use with Combifix VKS or SM Universal Shakers
Specifications
Sliding Plate | |
For SM30 with Combifix Edmund Bühler | |
2 Clamping Strips H, 2 Cramp Strips, 1 Spring Strip, additional base plate |