Learn More
Ibidi μ-Slide VI 0.4 Uncoated
Beschreibung
- A 6 channel μ-Slide suitable for flow experiments and for immunofluorescence assays
- bottom material: #1.5 Polymer Coverslip
- hydrophobic
- sterilized
Spezifikation
Spezifikation
| Type | Microscope Slide |
| Quantity | 15 |