Learn More
Ibidi μ-Slide VI 0.4 μ-Pattern RGD Test Patterns 3
Click to view available options
:
μ-Slide
Description
- A 6 channel slide with micropatterned surface to test different patterns for the establishment of cell culture assays
- bottom material: #1.5 Polymer Coverslip with passivated surface (biologically inert) and RGD pattern
- sterilized
Specifications
Specifications
| Type | Microscope Slide |
| Quantity | 2 |