Learn More
Ibidi μ-Slide VI 0.4 μ-Pattern RGD Test Patterns 3
Product Code. 17814794
Encompass_Preferred
Click to view available options
:
μ-Slide
Product Code. 17814794
Brand: Ibidi 83653
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
μ-Slide VI 0.4 μ-Pattern RGD Test Patterns 3 is a 6 channel slide with micropatterned surface.
  • A 6 channel slide with micropatterned surface to test different patterns for the establishment of cell culture assays
  • bottom material: #1.5 Polymer Coverslip with passivated surface (biologically inert) and RGD pattern
  • sterilized
Specifications
Type Microscope Slide
Quantity 2