missing translation for 'onlineSavingsMsg'
Learn More

ibidi μ-Slide Tissue Engineering ibiTreat

Product Code. 31941068
Encompass_Preferred
Change view
Click to view available options
No. per Case:
120/Cs.
15/Cs.
90/Cs.
No. per Pack:
1 Ea.
8/Tray, 15 Trays
Unit Size:
Pack of 120
Pack of 15
Pack of 90
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. per Case No. per Pack Unit Size
31941068 90/Cs. 1 Ea. Pack of 90
31941049 15/Cs. 1 Ea. Pack of 15
31941041 120/Cs. 8/Tray, 15 Trays Pack of 120
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 31941068 Supplier ibidi Supplier No. 8023690
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Microfluidic channel slide for tissue model and organ-on-a-chip applications

  • A μ-Slide for cell culture and high-end microscopy
  • No. 1.5 polymer coverslip
  • Tissue culture treated
  • Sterilized

Specifications

Type Chamber Slide
Surface Treatment ibiTreat
Material Polymer
For Use With (Application) Cell Culture, High-end Microscopy
No. per Case 90/Cs.
No. per Pack 1 Ea.
Sterility Sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.