Learn More
Ibidi μ-Slide Spheroid Perfusion ibiTreat
Product Code. 17246173
Encompass_Preferred
Click to view available options
:
μ-Slide
Product Code. 17246173
Brand: Ibidi 80356
Only null left
Add to basket
Add to basket
μ-Slide Spheroid Perfusion ibiTreat is a perfusable channel slide with 3 x 7 wells.
  • A perfusable channel slide with 3 x 7 wells for high-end microscopy
  • easy to connect with tubing and pump systems via female Luers
  • bottom material: #1.5 Polymer Coverslip
  • tissue culture treated
  • sterilized
Specifications
Type Microscope Slide
Quantity 15