Learn More
Ibidi μ-Slide Spheroid Perfusion ibiTreat
Click to view available options
:
μ-Slide
Description
- A perfusable channel slide with 3 x 7 wells for high-end microscopy
- easy to connect with tubing and pump systems via female Luers
- bottom material: #1.5 Polymer Coverslip
- tissue culture treated
- sterilized
Specifications
Specifications
| Type | Microscope Slide |
| Quantity | 15 |