Learn More
Ibidi μ-Slide III 3in1 Uncoated
Product Code. 17286163
Encompass_Preferred
Change view
Click to view available options
μ-Slide
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
17286163 μ-Slide
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17286163 Supplier Ibidi Supplier No. 80311
 Restricted Item
Estimated Shipment: 17-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
μ-Slide III 3in1 Uncoated is a μ-Slide for switchable chemotactic gradients.
  • A channel slide for switchable chemotactic gradients that merges 3 separated liquids into one channel
  • easy to connect with tubing and pump systems via female Luers
  • bottom material: #1.5 Polymer Coverslip
  • hydrophobic
  • sterilized
Specifications
Type Microscope Slide
Quantity 15