Learn More
Ibidi μ-Slide I 0.6 Luer Uncoated
Product Code. 16853983
Encompass_Preferred
Click to view available options
:
μ-Slide
Product Code. 16853983
Brand: Ibidi 80181
Please to purchase this item. Need a web account? Register with us today!
A μ-Slide has one channel for applying defined shear stress.
  • A μ-Slide with one channel for cell cultivation under flow and for applying defined shear stress
  • easy to connect with tubing and pump systems via female Luers
  • channel height: 600 μm
  • bottom material: #1.5 Polymer Coverslip
  • hydrophobic
  • sterilized
Specifications
Type Microscope Slide
Quantity 15