Learn More
Ibidi μ-Slide I 0.6 Luer Uncoated
Click to view available options
:
μ-Slide
Description
- A μ-Slide with one channel for cell cultivation under flow and for applying defined shear stress
- easy to connect with tubing and pump systems via female Luers
- channel height: 600 μm
- bottom material: #1.5 Polymer Coverslip
- hydrophobic
- sterilized
Specifications
Specifications
| Type | Microscope Slide |
| Quantity | 15 |