Learn More
Karl Hecht™ Assistent™ Antistatic Slide Cases
Click to view available options
Holds:
100 microslides
50 microslides
Unit Size:
Each
Description
- Stackable
- Black in color
- Resistant to xylene
- With numbered plastic grooves
Specifications
Specifications
| Product Type | Slide Box |
| Holds | 50 microslides |
| No. per Pack | 1 Ea. |