Learn More
Ibidi μ-Slide 8 Well High Glass Bottom, Bulk Pack
Product Code. 17266203
Encompass_Preferred
Change view
Click to view available options
:
μ-Slide
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
17266203 μ-Slide
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17266203 Supplier Ibidi Supplier No. 8080790
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
μ-Slide 8 Well High Glass Bottom, Bulk Pack is a chambered coverslip with 8 individual wells and high walls.
  • A chambered coverslip with 8 individual wells and high walls
  • suitable for use in TIRF and super-resolution microscopy
  • bottom material: #1.5H (170 μm ± 5 μm) D 263 M Schott glass coverslip
  • individually packed
  • sterilized
Specifications
Type Chambered Coverglass
Wells 8