Learn More
Ibidi μ-Slide 8 Well Glass Bottom, Bulk Pack
Description
- A chambered coverslip with 8 wells for use in TIRF and super-resolution microscopy
- bottom material: #1.5H (170 μm ± 5 μm) D 263 M Schott glass coverslip
- individually packed
- sterilized
Specifications
Specifications
| Type | Chambered Coverglass |
| Wells | 8 |
| Sterility | Sterile |