Learn More
Ibidi μ-Slide 2 Well Co-Culture ibiTreat
Product Code. 15573111
Encompass_Preferred
Click to view available options
:
μ-Slide
This item is not returnable. View return policy
Product Code. 15573111
Brand: Ibidi 81806
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy
μ-Slide 2 Well Co-Culture ibiTreat is a chambered coverslip with 2 x 9 wells.
  • A chambered coverslip with 2 x 9 wells suitable for co-cultivation assays in combination with microscopy
  • bottom material: #1.5 Polymer Coverslip
  • tissue culture treated
  • sterilized
Specifications
Type Chambered Coverglass
Wells 2
Sterility Sterile