Learn More
Ibidi μ-Slide 18 Well Uncoated
Product Code. 17206233
Encompass_Preferred
Click to view available options
:
μ-Slide
Product Code. 17206233
Brand: Ibidi 81811
Only null left
Add to basket
Add to basket
μ-Slide 18 Well Uncoated is a chambered coverslip with 18 wells.
  • A chambered coverslip with 18 wells for cell culture, immunofluorescence, and high-end microscopy
  • bottom material: #1.5 Polymer Coverslip
  • hydrophobic
  • sterilized
Specifications
Type Chambered Coverglass
Sterility Sterile