Learn More
Ibidi μ-Slide 15 Well 3D Uncoated
Description
- A chambered coverslip with 15 wells for 3D cell culture, tube formation assays, and immunofluorescence staining
- bottom material: #1.5 Polymer Coverslip
- hydrophobic
- sterilized
Specifications
Specifications
| Type | Coverglass |
| Sterility | Sterile |