Learn More
Ibidi μ-Slide 15 Well 3D Uncoated
Product Code. 17286223
Encompass_Preferred
Change view
Click to view available options
:
μ-Slide
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
17286223 μ-Slide
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17286223 Supplier Ibidi Supplier No. 81501
Please to purchase this item. Need a web account? Register with us today!
μ-Slide 15 Well 3D Uncoated is a chambered coverslip with 15 wells.
  • A chambered coverslip with 15 wells for 3D cell culture, tube formation assays, and immunofluorescence staining
  • bottom material: #1.5 Polymer Coverslip
  • hydrophobic
  • sterilized
Specifications
Type Coverglass
Sterility Sterile