missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLFN12 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marque: Invitrogen™ PA5116929
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Positive Samples: SH-SY5Y Immunogen sequence: DSWHVKDNRV MQLTRKEWIQ FMVEAEPKFS SSYEEVISQI NTSLPAPHSW PLLEWQRQRH HCPGLSGRIT Y.
Despite being first described several years ago, the roles of the Schlafen (SLFN) family of proteins remain largely unknown. The SLFN genes are preferentially expressed in lymphoid tissues and differentially regulated during thymocyte maturation. It is thought that many play roles in cell growth, hemopoietic cell differentiation, and T cell development and maturation. Most members contain at least one divergent AAA domain (AAA_4) that may play a role in ATP binding. Although also known as SLFN3, a Schlafen family member that may be a marker of T cell activation, human SLFN12 has relatively low homology to SLFN3 in other species. Loss of the SLFN12 gene due to deletion is associated with Kabuki syndrome, a multiple congenital anomaly syndrome, suggesting SLFN may play a role in this genetic condition.
Spécification
| SLFN12 | |
| Polyclonal | |
| Unconjugated | |
| SLFN12 | |
| Schlafen family member 12; SLFN12; SLFN3 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 55106 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.08 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q8IYM2 | |
| SLFN12 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 320-390 of human SLFN12 (NP_0012759381). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu