Learn More
Lenz Laborglasintrumente Sleeves with Grip Collar
Product Code. 11532344
Click to view available options
Standard Taper Joint No.:
NS 24/29
Unit Size:
Each
Product Code. 11532344
Brand: Lenz Laborglasintrumente 01101724
Only null left
Add to basket
Add to basket
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • Outstanding chemical resistance at operating temperatures from -200 to +260 °C
  • Sturdy design for permanent use
  • With grip collar and sealing rings for excellent tightness
Specifications
Material PTFE
Standard Taper Joint No. NS 24/29