missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC7A5 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
140.00 € - 399.00 €
Specifications
Antigen | SLC7A5 |
---|---|
Applications | Western Blot, Immunohistochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17561376
|
Proteintech
28670-1-AP-20UL |
20 μL |
140.00 €
20µL |
Estimated Shipment: 10-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17551376
|
Proteintech
28670-1-AP-150UL |
150 μL |
399.00 €
150µL |
Estimated Shipment: 10-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The SLC7A5 antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human SLC7A5. This antibody recognizes human antigen. The SLC7A5 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
SLC7A5 | |
Polyclonal | |
Rabbit | |
8140 | |
Primary |
Western Blot, Immunohistochemistry | |
Unconjugated | |
Human | |
IgG |