missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SLC6A7 Polyclonal Antibody
GREENER_CHOICE

Product Code. 18076141
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18076141 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18076141 Supplier Invitrogen™ Supplier No. PA5144911

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: REEGSLWERL QQASRPAMDW GPSLEENRTG MYVATLAGSQ SPKPLMVHMR KYGGITSFEN TAIEVDREIA Antigen sequence identity: Mouse 100%; Rat 99%.

This gene is a member of the gamma-aminobutyric acid (GABA) neurotransmitter gene family and encodes a high-affinity mammalian brain L-proline transporter protein. This transporter protein differs from other sodium-dependent plasma membrane carriers by its pharmacological specificity, kinetic properties, and ionic requirements.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC6A7
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SLC6A7
Gene Accession No. Q99884
Gene Alias AW455934; brain-specific L-proline transporter; L-proline transporter; proline transporter; Prot; Slc6a7; sodium-dependent proline transporter; solute carrier family 6 (neurotransmitter transporter), member 7; solute carrier family 6 (neurotransmitter transporter, L-proline), member 7; solute carrier family 6 member 7
Gene Symbols SLC6A7
Host Species Rabbit
Immunogen Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 560-629. Recombinant protein control fragment (Product #RP-110152).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6534
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.