missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC6A12 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA598160
This item is not returnable.
View return policy
Description
SLC6A12 Polyclonal Antibody for Western Blot, IHC (P), ELISA
Transports betaine and GABA. May have a role in regulation of GABAergic transmission in the brain through the reuptake of GABA into presynaptic terminals, as well as in osmotic regulation.
Specifications
| SLC6A12 | |
| Polyclonal | |
| Unconjugated | |
| SLC6A12 | |
| Betaine/GABA transporter 1; betaine/GABA transporter-1; BGT1; BGT-1; GABA transporter 1; Gabt2; gamma-aminobutyric acid (GABA-A) transporter 2; gamma-aminobutyric acid transporter; Gat1; GAT2; GAT-2; Na(+)/Cl(-) betaine/GABA transporter; RNU28927; SLC6A12; Sodium- and chloride-dependent betaine transporter; sodium- and chloride-dependent GABA transporter 2; solute carrier family 6 (neurotransmitter transporter), member 12; solute carrier family 6 (neurotransmitter transporter, betaine/GABA), member 12; solute carrier family 6 member 12; vesicular GABA transporter; VGAT | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 6539 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 0.82 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P48065 | |
| SLC6A12 | |
| Recombinant Human Sodium- and chloride-dependent betaine transporter protein (515-614AA). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction