Learn More
Invitrogen™ SLC34A2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5139878
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: APWPELGDAQ PNPDKYLEGA AGQQPTAPDK SKETNKNNTE APVTKIELLP Highest antigen sequence indentity to the following orthologs: Rat - 61%, Mouse - 59%.
SLC34A2 encodes a protein that is a pH-sensitive sodium-dependent phosphate transporter. Phosphate uptake is increased at lower pH. Defects in this gene are a cause of pulmonary alveolar microlithiasis. Three transcript variants encoding two different isoforms have been found for this gene.
Specifications
| SLC34A2 | |
| Polyclonal | |
| Unconjugated | |
| SLC34A2 | |
| AA536683; D5Ertd227e; Na(+)/Pi cotransporter 2B; Na(+)-dependent phosphate cotransporter 2B; NAPI IIb; NaPi-2b; NaPi3b; NAPI-3B; NAPI-IIb; Npt2b; NPTIIb; rNaPi IIb; Slc34a2; Sodium/phosphate cotransporter 2B; Sodium-dependent phosphate transport protein 2B; sodium-phosphate transport protein 2B; solute carrier family 34 (sodium phosphate), member 2; solute carrier family 34 (type II sodium/phosphate contransporter), member 2; solute carrier family 34 (type II sodium/phosphate cotransporter), member 2; solute carrier family 34 member 2; type II sodium-dependent phosphate transporter 3b; type IIb Na/Picotransporter; type IIb sodium-phosphate transporter | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10568 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O95436 | |
| SLC34A2 | |
| Recombinant protein corresponding to Human SLC34A2. Recombinant protein control fragment (Product #RP-109103). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.