missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SLC26A9 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15740212 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15740212 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15740212 Supplier Invitrogen™ Supplier No. PA562417

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: MSQPRPRYVV DRAAYSLTLF DDEFEKKDRT YPVGEKLRNA FRC Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 91%.

This gene is one member of a family of sulfate/anion transporter genes. Family members are well conserved in their genomic (number and size of exons) and protein (aa length among species) structures yet have markedly different tissue expression patterns.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC26A9
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SLC26A9
Gene Accession No. Q7LBE3
Gene Alias Anion transporter/exchanger protein 9; anion transporter/exchanger-9; E030002L01Rik; SLC26A9; SLC26A9 anion transporter/exchanger; solute carrier family 26 (anion exchanger), member 9; solute carrier family 26 member 9; solute carrier family 26, member 9
Gene Symbols SLC26A9
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SLC26A9. Recombinant protein control fragment (Product #RP-99802).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 115019
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.