missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC26A8 Polyclonal Antibody

Description
SLC26A8 Polyclonal Antibody for Western Blot, IHC (P)
SLC26A8 is one member of a family of sulfate/anion transporters. Family members are well conserved in their protein (aa length among species) structures yet have markedly different tissue expression patterns.
Specifications
Specifications
| Antigen | SLC26A8 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 |
| Gene | SLC26A8 |
| Gene Accession No. | Q96RN1 |
| Gene Alias | Anion exchange transporter; anion transporter/exchanger-8; RP11-482O9.1; SLC26A8; solute carrier family 26 (anion exchanger), member 8; solute carrier family 26 member 8; SPGF3; TAT1; testis anion transporter 1 |
| Gene Symbols | SLC26A8 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?