missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SLC22A3 Recombinant Rabbit Monoclonal Antibody (1K0A4)
GREENER_CHOICE

Product Code. 17296853 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17296853 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17296853 Supplier Invitrogen™ Supplier No. MA537983

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Positive Samples: PC-3, Mouse liver Immunogen sequence: GDKALQILRR IAKCNGKYLS SNYSEITVTD EEVSNPSFLD LVRTPQMRKC TLILMFAWFT SAVVYQGLVM RLGIIGGNLY IDFFISGVVE LPGALLILLT IERLGRRLPF AASNIVAGVA CLVTAFLPEG IAWLRTTVAT LGRLGITMAF.

Polyspecific organic cation transporters in the liver, kidney, intestine, and other organs are critical for elimination of many endogenous small organic cations as well as a wide array of drugs and environmental toxins. This gene is one of three similar cation transporter genes located in a cluster on chromosome 6. The encoded protein contains twelve putative transmembrane domains and is a plasma integral membrane protein. [provided by RefSeq, Jul 2008].
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC22A3
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone 1K0A4
Concentration 0.32 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene SLC22A3
Gene Accession No. O75751, Q9WTW5
Gene Alias 3-Oct; EMT; EMT organic cation transporter 3; EMTH; extraneuronal monoamine extraneuronal monoamine; extraneuronal monoamine transporter; Oct3; Orct3; Organic cation transporter 3; Slc22a3; Slca22a3; solute carrier family 22 (extraneuronal monoamine transporter), member 3; solute carrier family 22 (organic cation transporter), member 3; solute carrier family 22 member 3; solute carrier family 22, member 3
Gene Symbols SLC22A3
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20519, 6581
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.