missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC22A3 Antibody (SR2204), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22312-50ul
This item is not returnable.
View return policy
Description
SLC22A3 Monoclonal antibody specifically detects SLC22A3 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry
Specifications
| SLC22A3 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 6581 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| SR2204 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunohistochemistry 1:50-1:200 | |
| EMTEMTH, Extraneuronal monoamine transporter, OCT3EMT organic cation transporter 3, Organic cation transporter 3, solute carrier family 22 (extraneuronal monoamine transporter), member 3, solute carrier family 22 member 3 | |
| A synthesized peptide derived from human SLC22A3 (Uniprot #: O75751) | |
| 50 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction