missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SLC22A18 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15919634
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15919634 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15919634 Supplier Invitrogen™ Supplier No. PA566954

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: TQGLVIGQLSS HFSEEVLLR Highest antigen sequence identity to the following orthologs - mouse 75%, rat 70%.

ORCTL2 (organic cation transporter-like protein 2), also known as HET, ITM, BWR1A, IMPT1, TSSC5, BWSCR1A, SLC22A1L, p45-BWR1A or SLC22A18, is a 424 amino acid multi-pass membrane protein that localizes at the apical membrane surface of renal proximal tubules. ORCTL2 is expressed at high levels in the kidney, liver, colon and in fetal renal proximal tubules and present at lower levels in heart, brain and lung. Belonging to the major facilitator superfamily, ORCTL2 may act as a transporter of organic cations based on a proton efflux antiport mechanism and may also play a role in the transport of chloroquine and quinidine-related compounds in kidney. Defects in ORCTL2 are associated with breast and lung cancer and are the cause of rhabdomyosarcoma type 1, a malignant tumor derived from striated muscle.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC22A18
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SLC22A18
Gene Accession No. Q96BI1
Gene Alias AW260131; Beckwith-Wiedemann syndrome chromosomal region 1 candidate gene A protein; BWR1A; BWSCR1A; Efflux transporter-like protein; HET; imprinted multi-membrane spanning polyspecific transporter-related protein 1; imprinted multi-membrane-spanning polyspecific transporter-related protein 1; Impt1; ITM; Multi-membrane-spanning polyspecific transporter; ORCTL2; ORCTL-2; Organic cation transporter-like protein 2; organic cationic transporter-like 2; p45 Beckwith-Wiedemann region 1A; p45-Beckwith-Wiedemann region 1 A; p45-BWR1A; SLC22A18; SLC22A1L; solute carrier family 22 (organic cation transporter), member 18; Solute carrier family 22 member 18; Solute carrier family 22 member 1-like; solute carrier family 22, member 18; TSSC5; tumor suppressing subtransferable candidate 5; tumor-suppressing STF cDNA 5 protein; Tumor-suppressing subchromosomal transferable fragment candidate gene 5 protein
Gene Symbols SLC22A18
Host Species Rabbit
Immunogen Recombinant Human SLC22A18. Recombinant protein control fragment (Product #RP-107599).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5002
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.