Learn More
SLC22A10 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
483.00 €
Specifications
| Antigen | SLC22A10 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
SLC22A10 Polyclonal specifically detects SLC22A10 in Human samples. It is validated for Western Blot.Specifications
| SLC22A10 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| member 10, solute carrier family 22, member 10 | |
| SLC22A10 | |
| IgG |
| Western Blot | |
| Unconjugated | |
| RUO | |
| NP_001034841 | |
| 387775 | |
| Synthetic peptide directed towards the middle region of human SLC22A10. Peptide sequence QTLRVALACLGIGCSAATFSSVAVHFIELIPTVLRARASGIDLTASRIGA. | |
| Primary |