missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SKIDA1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA559179
This item is not returnable.
View return policy
Description
Immunogen sequence: GYLIIKGKQM FALSQVFTDL LKNIPRTTVH KRMDHLKVKK HHCDLEELRK LKAINSIAFH AAKCTLISRE DVEALYTSCK T Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 99%.
SKIDA1 is a protein coding gene.
Specifications
| SKIDA1 | |
| Polyclonal | |
| Unconjugated | |
| SKIDA1 | |
| 2810030E01Rik; 5730507N06Rik; BB088113; C10orf140; DLN1; DLN-1; LOW QUALITY PROTEIN: SKI/DACH domain-containing protein 1; protein DLN-1; SKI/DACH domain containing 1; SKI/DACH domain-containing protein 1; Skida1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 387640 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q1XH10 | |
| SKIDA1 | |
| Recombinant protein corresponding to Human SKIDA1. Recombinant protein control fragment (Product #RP-97179). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction