Learn More
SKI II, TRC
Specifications
Specifications
| Chemical Name or Material | SKI II |
| CAS | 312636-16-1 |
| Molecular Formula | C15H11ClN2OS |
| Quantity | 5 mg |
| Synonym | SKI 2, 4-[[4-(4-Chlorophenyl)-2-thiazolyl]amino]phenol, Sphingosine Kinase Inhibitor 2, SPHK I2 |
| InChI Formula | InChI=1S/C15H11ClN2OS/c16-11-3-1-10(2-4-11)14-9-20-15(18-14)17-12-5-7-13(19)8-6-12/h1-9,19H,(H,17,18) |
| SMILES | Oc1ccc(Nc2nc(cs2)c3ccc(Cl)cc3)cc1 |
| IUPAC Name | 4-[[4-(4-chlorophenyl)-1,3-thiazol-2-yl]amino]phenol |
| Molecular Weight (g/mol) | 302.78 |
| Formula Weight | 302.0281 |