missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ SJ 6986
Description
SJ 6986 is a potent and selective GSPT1/2 molecular glue Degrader (DC50 = 2.1 nM). The Degrader exhibits antiproliferative effects in human medulloblastoma and leukemia cell lines. (EC50 values are 0.4 nM, 1.5 nM, 336 nM, 726 nM and 3583 nM in MHH-CALL-4, MV4-11, MB004, MB002 and HD-MB03 cell lines, respectively). In vivo, SJ 6986 is stable in mouse (t1/2 = 4.7 h). This product is orally bioavailable.
Specifications
Specifications
| Chemical Name or Material | N-(2-(2,6-Dioxopiperidin-3-yl)-1,3-dioxoisoindolin-5-yl)-2-(trifluoromethoxy)benzenesulfonamide |
| CAS | 2765625-93-0 |
| Quantity | 10 mg |
| Target | Molecular Glues |
| Molecular Formula | C20H14F3N3O7S |
| Purity | 0.98 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?