missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ SIS17
Description
SIS17 is a selective HDAC11 inhibitor (IC50 = 0.83 μM). Inhibits demyristoylation of serine hydroxymethyl transferase 2 with no detectable activity at other HDAC subtypes. Combination with Oxaliplatin (Cat. No. 2623) increases cytotoxicity to K562 cells; improves survival of Oxaliplatin-treated 769P cells. Cell permeable.
Specifications
Specifications
| Chemical Name or Material | N'-Hexadecylthiophene-2-carbohydrazide |
| CAS | 2374313-54-7 |
| Quantity | 10 mg |
| Target | HDAC11 |
| Molecular Formula | C21H38N2OS |
| Purity | 0.95 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?