missing translation for 'onlineSavingsMsg'
Learn More

Sirtuin modulator 1, MedChemExpress

Product Code. 16430358
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16430358 5 mg 5mg
16497888 10 mg 10mg
16463837 50 mg 50mg
16437706 100 mg 100mg
16411859 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16430358 Supplier Medchem Express Supplier No. HY19758A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Sirtuin modulator 1 is a modulator of SIRTl, a homolog of SIRT3, with EC1.5 of < 1 μM, extracted from patent WO 2010071853 A1, Compound No.4.

Sirtuin modulator 1 is a modulator of SIRTl, a homolog of SIRT3, with EC1.5 of < 1 μM, extracted from patent WO 2010071853 A1, Compound No.4.

Specifications

Chemical Name or Material Sirtuin modulator 1
Physical Form Solid
CAS 2070015-26-6
Color Light Yellow
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C31H32ClN5O2S2
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : 12.5 mg/mL (20.62 mM; Need ultrasonic)
SMILES O=C(C1=C(CCCOC)SC(C2=CC=CC=C2)=N1)NC3=CC=CC=C3C4=NC5=C(N=CC(CN6CCCC6)=C5)S4.Cl[H]
Molecular Weight (g/mol) 606.2
Formula Weight 606.2
Percent Purity 99.72%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.