Learn More
Sipeimine, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
Description
Sipeimine is a natural product isolated from Fritillaria ussuriensis.
Specifications
Specifications
| Chemical Name or Material | Sipeimine |
| CAS | 61825-98-7 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C27H43NO3 |
| Quantity | 10 mg |
| Synonym | Imperialine |
| Solubility Information | DMSO : 8.33 mg/mL (19.39 mM; ultrasonic and warming and heat to 60°C) |
| SMILES | C[C@@]([C@@]1([H])C2=O)(CC[C@H](O)C1)[C@]3([H])[C@](C2)([H])[C@@](CC[C@]4([H])[C@@]5([H])CN(C[C@@H](C)CC6)[C@]6([H])[C@]4(O)C)([H])[C@@]5([H])C3 |
| Molecular Weight (g/mol) | 429.64 |
| Show More |