Learn More
Simvastatin Hydroxy Acid Sodium Salt, TRC
Product Code. 30004135
Change view
Click to view available options
Quantity:
1 mg
10 mg
25 mg
Unit Size:
10mg
1mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30004135 1 mg 1mg
30003584 25 mg 25mg
30010240 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30004135 Supplier Toronto Research Chemicals Supplier No. TRCS4850501MG
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 101314-97-0
Molecular Formula C25 H39 O6 . Na
Molecular Weight (g/mol) 458.56
Synonym 1-Naphthaleneheptanoic acid, 8-(2,2-dimethyl-1-oxobutoxy)-1,2,6,7,8,8a-hexahydro-β,δ-dihydroxy-2,6-dimethyl-, sodium salt (1:1), (βR,δR,1S,2S,6R,8S,8aR)-, 1-Naphthaleneheptanoic acid, 8-(2,2-dimethyl-1-oxobutoxy)-1,2,6,7,8,8a-hexahydro-β,δ-dihydroxy-2,6-dimethyl-, monosodium salt, (βR,δR,1S,2S,6R,8S,8aR)- (9CI), 1-Naphthaleneheptanoic acid, 8-(2,2-dimethyl-1-oxobutoxy)-1,2,6,7,8,8a-hexahydro-β,δ-dihydroxy-2,6-dimethyl-, monosodium salt, [1S-[1α(βS*,δS*),2α,6β,8β,8aα]]-, (3R,5R)-7-[(1S,2S,6R,8S,8aR)-8-[(2,2-Dimethylbutanoyl)oxy]-2,6-dimethyl-1,2,6,7,8,8a-hexahydronaphthalen-1-yl]-3,5-dihydroxyheptanoic Acid Sodium Salt, Simvastatin Imp. A (EP) as Sodium Salt, Tenivastatin Sodium Salt, Simvastatin Hydroxy Acid Sodium Salt
IUPAC Name sodium;(3R,5R)-7-[(1S,2S,6R,8S,8aR)-8-(2,2-dimethylbutanoyloxy)-2,6-dimethyl-1,2,6,7,8,8a-hexahydronaphthalen-1-yl]-3,5-dihydroxyheptanoate
SMILES [Na+].CCC(C)(C)C(=O)O[C@H]1C[C@@H](C)C=C2C=C[C@H](C)[C@H](CC[C@@H](O)C[C@@H](O)CC(=O)[O-])[C@@H]12
Specifications
Quantity 1 mg
InChI Formula InChI=1S/C25H40O6.Na/c1-6-25(4,5)24(30)31-21-12-15(2)11-17-8-7-16(3)20(23(17)21)10-9-18(26)13-19(27)14-22(28)29;/h7-8,11,15-16,18-21,23,26-27H,6,9-10,12-14H2,1-5H3,(H,28,29);/q;+1/p-1/t15-,16-,18+,19+,20-,21-,23-;/m0./s1
Formula Weight 458.2644
Percent Purity >95%
Chemical Name or Material (3R,5R)-7-[(1S,2S,6R,8S,8aR)-8-[(2,2-Dimethylbutanoyl)oxy]-2,6-dimethyl-1,2,6,7,8,8a-hexahydronaphthalen-1-yl]-3,5-dihydroxyheptanoic Acid Sodium Salt
Grade HPLC