Learn More
Macherey-Nagel™ Silylating Agent BSA
Product Code. 10125881 Shop All Macherey-Nagel Products
Change view
Click to view available options
Quantity:
10 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10125881 10 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10125881 Supplier Macherey-Nagel™ Supplier No. 701210.110
  • ONLINE EXCLUSIVE PRICE
    43.45 €
    42.63 € / Pack of 1
    Save 0.82 €
    1% Off
 Restricted Item
Estimated Shipment: 14-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Specifications
Item Description Silylating Agent BSA
Quantity 10 mL
compliance-icons compliance-icons compliance-icons
Product Identifier
  • BSA, 1x10 mL
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash hands thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: N,O-bis-trimethylsilyl-acetamide