missing translation for 'onlineSavingsMsg'
Learn More

Hanna Instruments™ Silver Reagent Sets, For Colorimetric Water Testing

Product Code. 10633852
Change view
Click to view available options
No. of Tests:
50
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Tests unitSize
10633852 50 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10633852 Supplier Hanna Instruments™ Supplier No. HI9373701

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Allows users to achieve fast and accurate colorimetric measurements. Hanna Instruments™ Silver Reagent Sets, For Colorimetric Water Testing are high-quality, pre-measured reagents for use with compatible benchtop and/or portable Hanna meters.

These high quality reagents are designed to follow an adaptation of the 1-(2-pyridylazo)-2-naphthol (PAN) method for colorimetric determination of silver. By simply adding the reagents to the sample as directed, a reaction will take place and a compatible meter can be used to determine the concentration from the orange color that is produced.

  • Pre-made, pre-measured water testing reagents for ease of use
  • Prepared with high purity chemicals manufactured in Hanna's state-of-the-art facility
  • Clearly marked with expiration date and lot number on each packet and bottle for traceability

Specifications

Analytes 1-(2-pyridylazo)-2-naphthol (PAN)
For Use With (Application) Silver determination
For Use With (Equipment) Compatible Hanna benchtop and portable photometers
Includes Dropper bottles
No. of Tests 50
Quantity 1 Set
Range 0.000 to 1.000mg/L silver
Test Method Cholorimetric determination
Type Water Testing Kit
compliance-icons compliance-icons
Product Identifier
  • Reagenzien für 50 Tests Silber
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H318-Causes serious eye damage.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P243-Take precautionary measures against static discharge.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P370+P378-In case of fire: Use powder to extinguish.
  • P403+P235-Store in a well-ventilated place. Keep cool.
Supplemental information
  • MIXTURE LIST-Contain: triton X-114, EDTA tetrasodium salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.