missing translation for 'onlineSavingsMsg'
Learn More

Silver nanoparticles, 100nm, 0.02 mg/ml, supplied in 2mM sodium citrate, 490nm absorption, Thermo Scientific Chemicals

Catalog Number p-7057652
Click to view available options
Quantity:
25 mL
100 mL
7440-22-4
Ag
107.87
MFCD00003397
BQCADISMDOOEFD-UHFFFAOYSA-N
argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
23954
CHEBI:9141
silver
[Ag]
7440-22-4
Ag
107.87
MFCD00003397
BQCADISMDOOEFD-UHFFFAOYSA-N
argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
23954
CHEBI:9141
silver
[Ag]

An alternative to gold nanoparticles due to lower absorption maximum in the 400-500nm range. Readily binds proteins and other ligands for conjugate and sensor development.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • An alternative to gold nanoparticles due to lower absorption maximum in the 400-500nm range
  • Readily binds proteins and other ligands for conjugate and sensor development
TRUSTED_SUSTAINABILITY
Color Yellow
Quantity 25 mL
Odor Odorless
Particle Size 100nm
Sensitivity Light sensitive
Form Liquid
Chemical Name or Material Silver nanoparticles, 0.02mg/mL, supplied in 2mM sodium citrate, 490nm absorption

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'