Learn More
Trajan Scientific and Medical™ SilTite™ Ferrules
Product Code. 15218923
Change view
Click to view available options
For Use With (Equipment):
Shimadzu Detector/Injectors, Standard MS Interface 0.32 mm I.D. Columns
Shimadzu Detector/Injectors, Standard MS Interface 0.45 to 0.53 mm I.D. Columns
Thermo GC/MS Interface Connections 0.10 to 0.25 mm I.D. Columns
Thermo GC/MS Interface Connections 0.32 mm I.D. Columns
Thermo GC/MS Interface Connections 0.45 to 0.53 mm I.D. Columns
Unit Size:
Pack of 10
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
15218923 Shimadzu Detector/Injectors, Standard MS Interface 0.32 mm I.D. Columns Pack of 10
15288923 Thermo GC/MS Interface Connections 0.45 to 0.53 mm I.D. Columns Pack of 10
15228923 Shimadzu Detector/Injectors, Standard MS Interface 0.45 to 0.53 mm I.D. Columns Pack of 10
15278923 Thermo GC/MS Interface Connections 0.32 mm I.D. Columns Pack of 10
15268923 Thermo GC/MS Interface Connections 0.10 to 0.25 mm I.D. Columns Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 15218923 Supplier Trajan Scientific and Medical™ Supplier No. 073228
Only null left
Add to basket
Add to basket
Ensures easy, leak-free installation for capillary columns without the use of any tools.
  • Eliminates leaks
  • No contamination
  • No need to retighten after temperature cycling
  • Cleaner spectra for better library matches
Specifications
Type SilTite Ferrules for 0.32 mm ID Columns for Shimadzu
For Use With (Equipment) Shimadzu Detector/Injectors, Standard MS Interface 0.32 mm I.D. Columns