Learn More
Hirschmann™ SILICOSEN™ Silicone Stoppers
Excellent for the cultivation of aerobic microorganisms. Specially processed culture plugs with continuous bubbles for preparation, filling and sterilization of culture media.
Brand: Hirschmann™ 8905720
Additional Details : Weight : 0.25000kg
Description
- Uniform pore structure
- Good air permeability
- Chemically and temperature resistant
- Liquid repellent and reusable
Specifications
SILICOSEN, C20 | |
Stopper | |
For The Preparation And Sterilization Of Culture Media | |
16 mm | |
Silicone | |
12 mm |
Natural | |
15-20 mm | |
20 mm | |
10/Pack | |
28 mm |