Learn More
Lauda™ Silicone tubes
Product Code. 10363772
Click to view available options
Diameter (Metric) Inside:
11 mm
Diameter (Metric) Well:
33 mm
Unit Size:
1m
Product Code. 10363772
Brand: Lauda™ LZS 007
Please to purchase this item. Need a web account? Register with us today!
Constructed from silicone. Lauda™ Silicone tubes are perfect for installation at higher air humidity condition.
  • For installation at higher air humidity
  • Made from silicone
Specifications
Temperature Range -70°C to +120°C
Type Tubing
Material EPDM Rubber
Diameter (Metric) Inside 11 mm
Diameter (Metric) Well 33 mm