Learn More
Hirschmann™ SILICOSEN™ Pink Silicone Stoppers
Product Code. 10660644
Click to view available options
Diameter (Metric) Inner:
6-9 mm
9-11 mm
11-14 mm
13-16 mm
15-18 mm
18-21 mm
20-23 mm
22-30 mm
30-40 mm
40-50 mm
Length (Metric):
10 mm
12 mm
13 mm
15 mm
20 mm
30 mm
35 mm
40 mm
Unit Size:
Pack of 10
Product Code. 10660644
Brand: Hirschmann™ 8905542
Only null left
Add to basket
Add to basket
Excellent for the cultivation of aerobic microorganisms. Hirschmann Instruments™ SILICOSEN™ Silicone Stoppers are specially processed culture plugs with continuous bubbles for preparation, filling and sterilization of culture media.
  • Uniform pore structure
  • Good air permeability
  • Chemically and temperature resistant
  • Liquid repellent and reusable
Specifications
Diameter (Metric) Inner 30-40 mm
For Use With (Application) For The Preparation And Sterilization Of Culture Media
Length (Metric) 35 mm
Quantity 10/Pk.
Color Pink
Material Silicone
Product Type Tapered Stopper
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis