Learn More
Hirschmann™ SILICOSEN™ Pink Silicone Stoppers
Product Code. 10402861
Click to view available options
Diameter (Metric) Inner:
6-9 mm
9-11 mm
11-14 mm
13-16 mm
15-18 mm
18-21 mm
20-23 mm
22-30 mm
30-40 mm
40-50 mm
Length (Metric):
10 mm
12 mm
13 mm
15 mm
20 mm
30 mm
35 mm
40 mm
Unit Size:
Pack of 10
Product Code. 10402861
Brand: Hirschmann™ 8905517
Only null left
Add to basket
Add to basket
Excellent for the cultivation of aerobic microorganisms. Hirschmann Instruments™ SILICOSEN™ Silicone Stoppers are specially processed culture plugs with continuous bubbles for preparation, filling and sterilization of culture media.
  • Uniform pore structure
  • Good air permeability
  • Chemically and temperature resistant
  • Liquid repellent and reusable
Specifications
Diameter (Metric) Inner 13-16 mm
For Use With (Application) For The Preparation And Sterilization Of Culture Media
Length (Metric) 10 mm
Quantity 10/Pk.
Color Pink
Material Silicone
Product Type Tapered Stopper