Learn More
JG Finneran Associates™ Silicone Septa
Product Code. 12472920
Encompass
Change view
Click to view available options
Closure Size:
10-425
13-425
8-425
Unit Size:
Pack of 1000
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Closure Size unitSize
12472920 13-425 Pack of 1000
12462920 10-425 Pack of 1000
10116980 10-425 Pack of 1000
12452920 8-425 Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 12472920 Supplier JG Finneran Associates™ Supplier No. 60406013
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
Specifications
Material (Liner) PTFE
Color Red
Material Silicone
Septum Yes
For Use With (Application) For Screw Thread Closures
Quantity 1000/Pk.
Closure Size 13-425