Learn More
Macherey-Nagel™ Silica N
75.40 € - 331.00 €
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10266811
|
Macherey-Nagel™
816330.5 |
5 kg |
331.00 €
5kg |
Estimated Shipment: 20-06-2024
Log in to see stock. |
|||||
10124811
|
Macherey-Nagel™
816330.1 |
1 kg |
75.40 €
1kg |
Estimated Shipment: 20-06-2024
Log in to see stock. |
|||||
Specifications
2 to 20 μm | |
Silica N | |
Silica |