Learn More
Hach™ Silica Low Range Reagent Set
Product Code. 10558583
Change view
Click to view available options
Range:
0.010 to 1.600 mg/L SiO2
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Range unitSize
10558583 0.010 to 1.600 mg/L SiO2 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10558583 Supplier Hach™ Supplier No. 2459300
Please to purchase this item. Need a web account? Register with us today!
For determination of Low Range Silica by the Heteropoly Blue method. Hach™ Silica Low Range Reagent Set includes Amino Acid F Reagent Powder Pillows, Citric Acid Powder Pillows and Molybdate 3 Reagent Solution for about 100 tests.
Powder Pillows make titration applications quick and easy.
  • Sealed packets ensure fresh, uncontaminated reagent every time
  • Premeasured to avoid measurement error and contamination from handling
  • Ready to use packets save time and hassle
  • Exact doses to easily monitor supply and prepare for lab procedures
  • Single-use packs for easy transportation and packing in test kits
TRUSTED_SUSTAINABILITY
Specifications
Description Powder pillows, Solution
For Use With (Application) Low range silica determination
For Use With (Equipment) Colorimeters (All Hach models except DR/3000, DR/2000, DR 900, PC II)
Includes Amino Acid F Reagent Powder Pillows, Citric Acid Powder Pillows and Molybdate 3 Reagent Solution
No. of Tests 100
Quantity 1 Set
Range 0.010 to 1.600 mg/L SiO2
Test Method Heteropoly blue
Type Regent set