Learn More
Hach™ Silica Low Range Reagent Set
Description
For determination of Low Range Silica by the Heteropoly Blue method. Hach™ Silica Low Range Reagent Set includes Amino Acid F Reagent Powder Pillows, Citric Acid Powder Pillows and Molybdate 3 Reagent Solution for about 100 tests.
Powder Pillows make titration applications quick and easy.
- Sealed packets ensure fresh, uncontaminated reagent every time
- Premeasured to avoid measurement error and contamination from handling
- Ready to use packets save time and hassle
- Exact doses to easily monitor supply and prepare for lab procedures
- Single-use packs for easy transportation and packing in test kits
Specifications
Specifications
| Description | Powder pillows, Solution |
| For Use With (Application) | Low range silica determination |
| For Use With (Equipment) | Colorimeters (All Hach models except DR/3000, DR/2000, DR 900, PC II) |
| Includes | Amino Acid F Reagent Powder Pillows, Citric Acid Powder Pillows and Molybdate 3 Reagent Solution |
| No. of Tests | 100 |
| Quantity | 1 Set |
| Range | 0.010 to 1.600 mg/L SiO2 |
| Test Method | Heteropoly blue |
| Type | Regent set |