Specifications
| Particle Size | 0.05 to 0.2 mm |
| Pore Size | ∼60 Å |
| Technique | Column Chromatography |
| Quantity | 1 kg |
| Surface Area (Metric) | ∼500 m2/g |
| Type | Silica 60 |
| For Use With (Application) | Low Pressure Column Chromatography |