missing translation for 'onlineSavingsMsg'
Learn More

Siglec-H Antibody (440c), Novus Biologicals™

Rat Monoclonal Antibody

Brand:  Novus Biologicals NBP3-11235

Product Code. 18314314

  • 486.00 € / 50µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Siglec-H Monoclonal specifically detects Siglec-H in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Functional.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Siglec-H
Monoclonal
0.1 mg/ml
Flow Cytometry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Functional
6430529G09Rik, Siglec-H, Siglech sialic acid binding Ig-like lectin H
The monoclonal antibody 440c reacts with Siglec-H, a cell-surface receptor molecule selectively expressed on murine natural interferon producing cells (IPC), also called mouse plasmacytoid dendritic cells (pDC).
50 μg
Primary
Mouse
Purified
Flow Cytometry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Functional Assay
440c
Unconjugated
0.2 um filtered solution in PBS, 0.1% BSA
Rat
Protein G purified
RUO
233274
Store at 4°C. Do not freeze.
IgG2b
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.